siRNA Details
| Gene Symbol: ERLEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127397.2 | NP_001120869.1 | 2, 3 | 555 | ||
| NM_001127398.2 | NP_001120870.1 | 2, 3 | 555 | |||
| NM_015701.4 | NP_056516.2 | 2, 3 | 555 | |||
| XM_011532766.1 | XP_011531068.1 | 2, 3 | 469 | |||
| GenBank mRNA | AK001913.1 | - | 2, 3 | 436 | ||
| AK315477.1 | - | 2, 3 | 344 | |||
| AY358717.1 | - | 2, 3 | 439 | |||
| AY453410.1 | - | 2, 3 | 406 | |||
| BC013129.1 | - | 2, 3 | 406 | 3447840 | ||
| BC022228.1 | - | 2, 3 | 403 | 4447432 | ||
| EU794676.1 | - | 2, 3 | 555 | |||

ERLEC1
Genomic Map