siRNA Details
| Gene Symbol: ZNF593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015871.4 | NP_056955.2 | 1, 2 | 298 | ||
| XM_017001398.1 | XP_016856887.1 | 1, 2 | 667 | |||
| GenBank mRNA | AK311926.1 | - | 1, 2 | 298 | ||
| BC002580.2 | - | 1, 2 | 276 | 3143091 | ||
| BC019267.1 | - | 1, 2 | 271 | 2823658 | ||
| D45213.1 | - | 1, 2 | 207 | |||
| DA124660.1 | - | 1, 2 | 298 | |||

ZNF593
Genomic Map