siRNA Details
| Gene Symbol: PI15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324403.1 | NP_001311332.1 | 5, 6 | 857 | ||
| NM_015886.4 | NP_056970.1 | 5, 6 | 829 | |||
| GenBank mRNA | BC074931.2 | - | 4, 5 | 673 | 30915489 | |
| BC074932.2 | - | 4, 5 | 673 | 30915266 | ||
| BC126290.1 | - | 4, 5 | 678 | 8992006 | ||
| BC126292.1 | - | 4, 5 | 680 | 8992008 | ||
| CR749657.1 | - | 5, 6 | 830 | |||
| D45027.1 | - | 4, 5 | 1134 | |||

PI15
Genomic Map