siRNA Details
| Gene Symbol: EPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015630.3 | NP_056445.3 | 7 | 1013 | ||
| XM_011510941.2 | XP_011509243.1 | 7 | 1224 | |||
| XM_011510943.2 | XP_011509245.1 | 6 | 897 | |||
| GenBank mRNA | AF286904.1 | - | 7 | 1005 | ||
| AK001433.1 | - | 5 | 604 | |||
| AK125789.1 | - | 8 | 1082 | |||
| BC093818.1 | - | 7 | 925 | 7939663 | ||
| BC093820.1 | - | 7 | 925 | 7939665 | ||

EPC2
Genomic Map