siRNA Details
| Gene Symbol: KIF1BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015634.3 | NP_056449.1 | 2 | 597 | ||
| GenBank mRNA | AB033105.1 | - | 2 | 520 | ||
| AK291343.1 | - | 2 | 502 | |||
| BC006427.1 | - | 2 | 520 | |||
| BC012180.1 | - | 2 | 494 | 4550085 | ||

KIF1BP
Genomic Map