siRNA Details
| Gene Symbol: IRF2BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015649.2 | NP_056464.1 | 1 | 2457 | ||
| GenBank mRNA | AK092407.1 | - | 1 | 1712 | ||
| AL080159.1 | - | 1 | 1394 | |||
| AY278022.1 | - | 1 | 2408 | |||
| BC005834.2 | - | 1 | 1845 | |||
| BC038222.1 | - | 1 | 2405 | 5168442 | ||
| BC039002.1 | - | 1 | 2422 | 5182566 | ||
| BC078181.1 | - | 1 | 2130 | 6501537 | ||

IRF2BP1
Genomic Map