siRNA Details
| Gene Symbol: C10orf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015652.2 | NP_056467.2 | 1 | 235 | ||
| GenBank mRNA | AK126886.1 | - | 1 | 288 | ||
| BC024315.1 | - | 1 | 235 | 3903728 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf12| Gene Symbol: C10orf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015652.2 | NP_056467.2 | 1 | 235 | ||
| GenBank mRNA | AK126886.1 | - | 1 | 288 | ||
| BC024315.1 | - | 1 | 235 | 3903728 | ||