siRNA Details
| Gene Symbol: NOC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015658.3 | NP_056473.2 | 6 | 745 | ||
| GenBank mRNA | AK024284.1 | - | 6 | 699 | ||
| AK225239.1 | - | 6 | 700 | |||
| AK315080.1 | - | 6 | 714 | |||
| AL050019.1 | - | 6 | 701 | |||
| BC003555.1 | - | 6 | 718 | 3546209 | ||

NOC2L
Genomic Map