siRNA Details
| Gene Symbol: HMGCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166107.1 | NP_001159579.1 | 1, 2 | 200 | ||
| NM_005518.3 | NP_005509.1 | 1, 2 | 200 | |||
| XM_011541313.1 | XP_011539615.1 | 1, 2 | 393 | |||
| GenBank mRNA | AK301594.1 | - | 1, 2 | 170 | ||
| AK302276.1 | - | 1, 2 | 173 | |||
| AK303777.1 | - | 1, 2 | 173 | |||
| AK313590.1 | - | 1, 2 | 175 | |||
| BC044217.1 | - | 1, 2 | 153 | 5744956 | ||
| BF964973.1 | - | 1, 2 | 174 | |||
| BP420631.1 | - | 1, 2 | 209 | |||
| CR456850.1 | - | 1, 2 | 112 | |||
| X83618.1 | - | 1, 2 | 163 | |||

HMGCS2
Genomic Map