siRNA Details
| Gene Symbol: G0S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015714.3 | NP_056529.1 | 2 | 780 | ||
| GenBank mRNA | AV709160.1 | - | 1 | 195 | ||
| BC009694.1 | - | 2 | 684 | 3891372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G0S2| Gene Symbol: G0S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015714.3 | NP_056529.1 | 2 | 780 | ||
| GenBank mRNA | AV709160.1 | - | 1 | 195 | ||
| BC009694.1 | - | 2 | 684 | 3891372 | ||