siRNA Details
| Gene Symbol: CDC40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015891.2 | NP_056975.1 | 1, 2 | 221 | ||
| GenBank mRNA | AB208903.1 | - | 1, 2 | 195 | ||
| AF015044.1 | - | 1, 2 | 188 | |||
| AF038392.1 | - | 1, 2 | 221 | |||
| AF061241.1 | - | 1, 2 | 194 | |||
| AK222721.1 | - | 1, 2 | 218 | |||
| AK295055.1 | - | 1, 2 | 214 | |||
| AK314601.1 | - | 1, 2 | 217 | |||
| BC111061.1 | - | 2, 3 | 298 | |||
| BC117145.1 | - | 1, 2 | 218 | 40125696 | ||
| BC126114.1 | - | 1, 2 | 218 | 8991830 | ||

CDC40
Genomic Map