siRNA Details
| Gene Symbol: ERLEC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127397.2 | NP_001120869.1 | 13 | 1766 | ||
| NM_001127398.2 | NP_001120870.1 | 13 | 1682 | |||
| NM_015701.4 | NP_056516.2 | 14 | 1844 | |||
| GenBank mRNA | AF131743.1 | - | 6 | 663 | ||
| AF131849.1 | - | 8 | 1033 | |||
| AK001913.1 | - | 14 | 1725 | |||
| AY358717.1 | - | 13 | 1566 | |||
| AY453410.1 | - | 14 | 1695 | |||
| BC013129.1 | - | 14 | 1695 | 3447840 | ||
| BC022228.1 | - | 13 | 1530 | 4447432 | ||
| EU794676.1 | - | 14 | 1844 | |||

ERLEC1
Genomic Map