siRNA Details
| Gene Symbol: ZNF593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015871.4 | NP_056955.2 | 3 | 640 | ||
| GenBank mRNA | BC019267.1 | - | 3 | 613 | 2823658 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF593| Gene Symbol: ZNF593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015871.4 | NP_056955.2 | 3 | 640 | ||
| GenBank mRNA | BC019267.1 | - | 3 | 613 | 2823658 | |