siRNA Details
| Gene Symbol: POMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015932.5 | NP_057016.1 | 4 | 402 | ||
| GenBank mRNA | AF077200.1 | - | 4 | 303 | ||
| AF125097.1 | - | 4 | 302 | |||
| AF262975.1 | - | 4 | 246 | |||
| AF275807.1 | - | 4 | 251 | |||
| AK312118.1 | - | 4 | 273 | |||
| BC003390.1 | - | 4 | 265 | 3446986 | ||
| BC014334.1 | - | 4 | 296 | 4045091 | ||
| EF535527.1 | - | 4 | 234 | |||
| GU045555.1 | - | 4 | 402 | |||
| GU045556.1 | - | 4 | 401 | |||

POMP
Genomic Map