siRNA Details
| Gene Symbol: HEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015987.4 | NP_057071.2 | 1, 2 | 281 | ||
| GenBank mRNA | AF117615.1 | - | 1, 2 | 164 | ||
| AF167473.1 | - | 1, 2 | 86 | |||
| AK289877.1 | - | 1, 2 | 245 | |||
| BC016277.2 | - | 1, 2 | 116 | 3907035 | ||
| BI550915.1 | - | 1, 2 | 285 | |||
| BT007294.1 | - | 1, 2 | 86 | |||
| CF126276.1 | - | 1, 2 | 282 | |||

HEBP1
Genomic Map