siRNA Details
| Gene Symbol: UTP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016001.2 | NP_057085.2 | 1, 2 | 398 | ||
| XM_006721930.3 | XP_006721993.2 | 1, 2 | 377 | |||
| XM_011524870.2 | XP_011523172.1 | 1, 2 | 377 | |||
| GenBank mRNA | AF151806.1 | - | 1, 2 | 384 | ||
| AK026355.1 | - | 2, 3 | 147 | |||
| AK314399.1 | - | 1, 2 | 377 | |||
| BC025276.1 | - | 1, 2 | 374 | 4861495 | ||
| BG723023.1 | - | 1, 2 | 410 | |||

UTP18
Genomic Map