siRNA Details
| Gene Symbol: ZDHHC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008222.2 | NP_001008223.1 | 2 | 497 | ||
| NM_016032.3 | NP_057116.2 | 3 | 477 | |||
| XM_011531348.2 | XP_011529650.1 | 3 | 464 | |||
| GenBank mRNA | AB209807.1 | - | 2 | 416 | ||
| AF151847.1 | - | 3 | 365 | |||
| AK001112.1 | - | 3 | 427 | |||
| AK001524.1 | - | 2 | 228 | |||
| AL161962.1 | - | 3 | 354 | |||
| AY358558.1 | - | 3 | 409 | |||
| AY952881.8 | - | 3 | 449 | |||
| BC000035.2 | - | 2 | 371 | 3506601 | ||
| BC003128.1 | - | 3 | 415 | 2957911 | ||
| BC006200.2 | - | 3 | 411 | 2964425 | ||
| BC012826.2 | - | 3 | 411 | 2964425 | ||
| BM909699.1 | - | 404 | ||||
| CD627424.1 | - | 3 | 442 | |||
| DA493628.1 | - | 3 | 477 | |||
| DA768650.1 | - | 2 | 497 | |||

ZDHHC9
Genomic Map