siRNA Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016047.3 | NP_057131.1 | 3 | 395 | ||
| GenBank mRNA | AF151868.1 | - | 3 | 391 | ||
| AF161523.1 | - | 3 | 289 | |||
| AF184213.1 | - | 3 | 391 | |||
| AF401310.1 | - | 3 | 180 | |||
| AK000621.1 | - | 3 | 295 | |||
| AK311118.1 | - | 2 | 182 | |||
| AK311908.1 | - | 3 | 328 | |||
| BC015463.1 | - | 3 | 395 | 3914184 | ||
| BG200650.1 | - | 2 | 156 | |||

SF3B6
Genomic Map