siRNA Details
| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015920.3 | NP_057004.1 | 3, 4 | 371 | ||
| GenBank mRNA | AF070668.1 | - | 3, 4 | 293 | ||
| AK024591.1 | - | 3, 4 | 306 | |||
| BC003667.1 | - | 3, 4 | 303 | 3827362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPS27L| Gene Symbol: RPS27L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015920.3 | NP_057004.1 | 3, 4 | 371 | ||
| GenBank mRNA | AF070668.1 | - | 3, 4 | 293 | ||
| AK024591.1 | - | 3, 4 | 306 | |||
| BC003667.1 | - | 3, 4 | 303 | 3827362 | ||