siRNA Details
| Gene Symbol: PELO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015946.4 | NP_057030.3 | 2 | 1131 | ||
| GenBank mRNA | AF132951.1 | - | 1 | 405 | ||
| AF139828.1 | - | 1 | 378 | |||
| AK025729.1 | - | 1 | 445 | |||
| AK056144.1 | - | 2 | 1131 | |||
| AK312633.1 | - | 1 | 383 | |||
| AY117399.1 | - | 2 | 1131 | |||
| BC005889.2 | - | 1 | 419 | 2967612 | ||
| BC007249.2 | - | 1 | 419 | 2967002 | ||
| BC007650.1 | - | 1 | 436 | 3608639 | ||
| BC022789.1 | - | 1 | 330 | 3160699 | ||

PELO
Genomic Map