siRNA Details
| Gene Symbol: SLC35B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142541.2 | NP_001136013.1 | 4, 5 | 585 | ||
| NM_015948.4 | NP_057032.2 | 4, 5 | 947 | |||
| NR_109913.1 | 4, 5 | 796 | ||||
| XM_005249156.3 | XP_005249213.1 | 3, 4 | 727 | |||
| XM_005249157.2 | XP_005249214.1 | 4, 5 | 720 | |||
| XM_005249158.1 | XP_005249215.1 | 3, 4 | 615 | |||
| XM_006715102.3 | XP_006715165.1 | 4, 5 | 757 | |||
| XM_006715103.3 | XP_006715166.1 | 3, 4 | 727 | |||
| XM_011514648.1 | XP_011512950.1 | 5, 6 | 727 | |||
| XM_011514650.2 | XP_011512952.1 | 3, 4 | 727 | |||
| XM_017010910.1 | XP_016866399.1 | 4, 5 | 1043 | |||
| XM_017010911.1 | XP_016866400.1 | 3, 4 | 961 | |||
| XM_017010912.1 | XP_016866401.1 | 4, 5 | 675 | |||
| XM_017010913.1 | XP_016866402.1 | 4, 5 | 1043 | |||
| XM_017010914.1 | XP_016866403.1 | 3, 4 | 811 | |||
| XM_017010915.1 | XP_016866404.1 | 3, 4 | 615 | |||
| GenBank mRNA | AB231931.1 | - | 2, 3 | 422 | ||
| AF132953.1 | - | 3, 4 | 443 | |||
| AK313522.1 | - | 4, 5 | 563 | |||
| BC006973.1 | - | 4, 5 | 773 | 3827889 | ||
| BX538271.1 | - | 4, 5 | 791 | |||
| BX648022.1 | - | 3, 4 | 4884 | |||

SLC35B3
Genomic Map