siRNA Details
| Gene Symbol: RWDD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007464.2 | NP_001007465.1 | 4, 5 | 608 | ||
| NM_015952.3 | NP_057036.2 | 3, 4 | 494 | |||
| NM_016104.3 | NP_057188.2 | 5, 6 | 736 | |||
| GenBank mRNA | AF092134.1 | - | 3, 4 | 451 | ||
| AF132958.1 | - | 3, 4 | 364 | |||
| AK290727.1 | - | 3, 4 | 494 | |||
| AK301089.1 | - | 4, 5 | 471 | |||
| BC015802.1 | - | 3, 4 | 350 | 3909348 | ||
| CF128570.1 | - | 3, 4 | 441 | |||
| CN363729.1 | - | 5, 6 | 577 | |||

RWDD1
Genomic Map