siRNA Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015991.2 | NP_057075.1 | 3 | 286 | ||
| GenBank mRNA | AF135157.1 | - | 2 | 213 | ||
| AK311980.1 | - | 3 | 229 | |||
| BC030153.2 | - | 3 | 256 | 4850418 | ||
| BC071986.1 | - | 2 | 222 | 6300836 | ||
| BG548439.1 | - | 3 | 287 | |||

C1QA
Genomic Map