siRNA Details
| Gene Symbol: SF3B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016047.3 | NP_057131.1 | 3 | 418 | ||
| GenBank mRNA | AF151868.1 | - | 3 | 414 | ||
| AF161523.1 | - | 3 | 312 | |||
| AF184213.1 | - | 3 | 414 | |||
| AF401310.1 | - | 3 | 203 | |||
| AK000621.1 | - | 3 | 318 | |||
| AK311118.1 | - | 2 | 205 | |||
| AK311908.1 | - | 3 | 351 | |||
| BC015463.1 | - | 3 | 418 | 3914184 | ||
| BG200650.1 | - | 2 | 179 | |||

SF3B6
Genomic Map