siRNA Details
| Gene Symbol: POP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015918.3 | NP_057002.2 | 4 | 396 | ||
| NM_198202.1 | NP_937845.1 | 3 | 250 | |||
| XM_011538441.1 | XP_011536743.1 | 4 | 508 | |||
| GenBank mRNA | AF070660.1 | - | 4 | 380 | ||
| AF117232.1 | - | 4 | 380 | |||
| AJ306296.1 | - | 4 | 383 | |||
| AK223206.1 | - | 4 | 368 | |||
| AK303144.1 | - | 3 | 231 | |||
| BC012505.1 | - | 4 | 383 | 4708484 | ||
| BI668578.1 | - | 4 | 403 | |||
| BU155588.1 | - | 239 | ||||
| CB995722.1 | - | 4 | 518 | |||

POP5
Genomic Map