siRNA Details
| Gene Symbol: CUTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014433.2 | NP_001014433.1 | 6 | 609 | ||
| NM_001014837.1 | NP_001014837.1 | 6 | 858 | |||
| NM_001014838.1 | NP_001014838.1 | 6 | 827 | |||
| NM_001014840.1 | NP_001014840.1 | 6 | 737 | |||
| NM_015921.2 | NP_057005.1 | 5 | 1000 | |||
| GenBank mRNA | AF106943.1 | - | 5 | 637 | ||
| AF230924.1 | - | 5 | 423 | |||
| AK311556.1 | - | 1 | 1367 | |||
| BC005890.2 | - | 6 | 434 | 3505574 | ||
| BC107751.1 | - | 5 | 637 | 6714433 | ||

CUTA
Genomic Map