siRNA Details
| Gene Symbol: POMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015932.5 | NP_057016.1 | 6 | 799 | ||
| GenBank mRNA | BC003390.1 | - | 6 | 662 | 3446986 | |
| BC014334.1 | - | 6 | 693 | 4045091 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POMP| Gene Symbol: POMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015932.5 | NP_057016.1 | 6 | 799 | ||
| GenBank mRNA | BC003390.1 | - | 6 | 662 | 3446986 | |
| BC014334.1 | - | 6 | 693 | 4045091 | ||