siRNA Details
| Gene Symbol: DERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304777.1 | NP_001291706.1 | 7 | 1150 | ||
| NM_001304779.1 | NP_001291708.1 | 4 | 789 | |||
| NM_016041.4 | NP_057125.2 | 7 | 1153 | |||
| NR_130905.1 | 6 | 1265 | ||||
| NR_130906.1 | 6 | 1059 | ||||
| GenBank mRNA | AF151859.1 | - | 7 | 1091 | ||
| AF208065.1 | - | 7 | 1089 | |||
| AF242523.1 | - | 7 | 1089 | |||
| AK024574.1 | - | 3 | 771 | |||
| BC010890.1 | - | 7 | 1101 | 4280273 | ||

DERL2
Genomic Map