siRNA Details
| Gene Symbol: EMC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016049.3 | NP_057133.2 | 5, 6 | 600 | ||
| XM_005267721.4 | XP_005267778.1 | 4, 5 | 1568 | |||
| GenBank mRNA | AF151870.1 | - | 5, 6 | 595 | ||
| AY368908.1 | - | 5, 6 | 545 | |||
| BC002491.2 | - | 5, 6 | 600 | 3051105 | ||

EMC9
Genomic Map