siRNA Details
| Gene Symbol: MRPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016065.3 | NP_057149.1 | 2 | 434 | ||
| XM_017016305.1 | XP_016871794.1 | 2 | 429 | |||
| GenBank mRNA | AB049948.1 | - | 2 | 320 | ||
| AF151890.1 | - | 2 | 387 | |||
| AK025715.1 | - | 2 | 367 | |||
| AK098291.1 | - | 2 | 380 | |||
| BC021106.1 | - | 2 | 329 | 5020046 | ||

MRPS16
Genomic Map