siRNA Details
| Gene Symbol: SAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033503.2 | NP_001028675.1 | 4 | 370 | ||
| NM_016103.3 | NP_057187.1 | 3 | 251 | |||
| XM_017009521.1 | XP_016865010.1 | 3 | 258 | |||
| GenBank mRNA | AF087850.1 | - | 3 | 288 | ||
| AF092130.1 | - | 3 | 267 | |||
| AK223452.1 | - | 3 | 236 | |||
| AL710660.1 | - | 4 | 352 | |||
| BC002847.2 | - | 4 | 301 | 3636540 | ||
| BC093034.1 | - | 3 | 251 | 30528518 | ||
| CD048852.1 | - | 4 | 378 | |||

SAR1B
Genomic Map