siRNA Details
| Gene Symbol: TELO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016111.3 | NP_057195.2 | 6 | 1181 | ||
| XM_011522773.2 | XP_011521075.1 | 6 | 1172 | |||
| XM_011522774.2 | XP_011521076.1 | 6 | 1370 | |||
| XM_011522775.2 | XP_011521077.1 | 6 | 1153 | |||
| XM_011522776.2 | XP_011521078.1 | 6 | 1240 | |||
| XM_011522777.2 | XP_011521079.1 | 6 | 1172 | |||
| XM_011522778.2 | XP_011521080.1 | 6 | 1172 | |||
| XM_017023914.1 | XP_016879403.1 | 6 | 1153 | |||
| GenBank mRNA | AB014583.3 | - | 6 | 1169 | ||
| AK302103.1 | - | 6 | 919 | |||
| AL080126.1 | - | 6 | 1112 | |||
| AL137394.1 | - | 6 | 1112 | |||
| BC017188.2 | - | 5 | 937 | 3139743 | ||

TELO2
Genomic Map