siRNA Details
Gene Symbol: GPR75-ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164165.1 | NP_001157637.1 | 3 | 462 | ||
GenBank mRNA | BC110915.1 | - | 3 | 384 | 5764788 |
Gene Symbol: ASB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201965.1 | NP_001188894.1 | 2 | 611 | ||
NM_016115.4 | NP_057199.1 | 3 | 445 | |||
NM_145863.2 | NP_665862.1 | 2 | 236 | |||
GenBank mRNA | AB209795.1 | - | 3 | 337 | ||
AF156778.1 | - | 2 | 243 | |||
AK000985.1 | - | 3 | 371 | |||
AK001283.1 | - | 2 | 169 | |||
AK056069.1 | - | 2 | 613 | |||
AK222595.1 | - | 2 | 255 | |||
AK222715.1 | - | 2 | 227 | |||
AK303133.1 | - | 3 | 371 | |||
BC006488.2 | - | 2 | 257 | 3537359 | ||
BC009569.1 | - | 3 | 303 | 4052577 | ||
DA038561.1 | - | 2 | 236 |
