siRNA Details
| Gene Symbol: UBAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016172.2 | NP_057256.2 | 2 | 453 | ||
| GenBank mRNA | AF038200.1 | - | 1 | 18 | ||
| AF068195.1 | - | 1 | 80 | |||
| AF176796.1 | - | 2 | 426 | |||
| BC004967.1 | - | 2 | 453 | 3535682 | ||
| BC011822.1 | - | 2 | 279 | 3688526 | ||

UBAC1
Genomic Map