siRNA Details
| Gene Symbol: HN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002032.2 | NP_001002032.1 | 2 | 367 | ||
| NM_001002033.2 | NP_001002033.1 | 3 | 462 | |||
| NM_001288609.1 | NP_001275538.1 | 2 | 1051 | |||
| NM_001288610.1 | NP_001275539.1 | 2 | 414 | |||
| NM_001288611.1 | NP_001275540.1 | 2 | 367 | |||
| NM_016185.3 | NP_057269.1 | 2 | 367 | |||
| NR_109933.1 | 2 | 367 | ||||
| GenBank mRNA | AF086910.2 | - | 2 | 367 | ||
| AF177862.1 | - | 2 | 184 | |||
| AF266846.1 | - | 2 | 196 | |||
| AK308253.1 | - | 2 | 414 | |||
| AK312613.1 | - | 2 | 196 | |||
| AY322169.1 | - | 2 | 359 | |||
| AY322170.1 | - | 3 | 454 | |||
| BC001420.2 | - | 2 | 185 | 3140086 | ||
| BC039343.1 | - | 2 | 1053 | 5269120 | ||
| BG779109.1 | - | 2 | 197 | |||
| CD359142.1 | - | 363 | ||||
| DA604735.1 | - | 2 | 196 | |||
| DB451153.1 | - | 2 | 383 | |||

HN1
Genomic Map