siRNA Details
| Gene Symbol: CCDC53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301107.1 | NP_001288036.1 | 4 | 402 | ||
| NM_016053.3 | NP_057137.1 | 4 | 402 | |||
| XM_011538423.1 | XP_011536725.1 | 4 | 320 | |||
| XM_017019383.1 | XP_016874872.1 | 3 | 217 | |||
| GenBank mRNA | AF151874.1 | - | 4 | 347 | ||
| AF155655.1 | - | 4 | 347 | |||
| AK314970.1 | - | 4 | 320 | |||
| BC010889.1 | - | 4 | 273 | 4272385 | ||
| BX356987.2 | - | 4 | 357 | |||
| CR457171.1 | - | 4 | 240 | |||
| DB452335.1 | - | 4 | 402 | |||

CCDC53
Genomic Map