siRNA Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016063.2 | NP_057147.2 | 5, 6 | 729 | ||
| GenBank mRNA | AF151888.1 | - | 4, 5 | 486 | ||
| AF529363.1 | - | 5, 6 | 525 | |||
| BC001671.2 | - | 4, 5 | 561 | |||
| BC003357.1 | - | 5, 6 | 568 | |||
| BC066332.1 | - | 6, 7 | 543 | 5294646 | ||
| BE743302.1 | - | 5, 6 | 642 | |||
| BP257644.1 | - | 1, 2 | 116 | |||

HDDC2
Genomic Map