siRNA Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317980.1 | NP_001304909.1 | 3 | 294 | ||
| NM_001317981.1 | NP_001304910.1 | 3 | 268 | |||
| NM_001317982.1 | NP_001304911.1 | 3 | 250 | |||
| NM_016093.3 | NP_057177.1 | 3 | 240 | |||
| XM_005265921.3 | XP_005265978.1 | 3 | 291 | |||
| XM_011534565.2 | XP_011532867.1 | 3 | 855 | |||
| XM_017009519.1 | XP_016865008.1 | 3 | 282 | |||
| XM_017009520.1 | XP_016865009.1 | 3 | 213 | |||
| GenBank mRNA | AF083248.1 | - | 3 | 214 | ||
| AK128872.1 | - | 1 | 208 | |||
| BC017360.2 | - | 3 | 228 | 4551093 | ||
| BC070192.1 | - | 3 | 213 | 6727802 | ||
| BI838595.1 | - | 268 | ||||
| BQ278240.1 | - | 3 | 241 | |||
| BU193755.1 | - | 3 | 274 | |||
| BU569345.1 | - | 3 | 253 | |||

RPL26L1
Genomic Map