siRNA Details
| Gene Symbol: MPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270879.1 | NP_001257808.1 | 6 | 712 | ||
| NM_016098.3 | NP_057182.1 | 5 | 496 | |||
| NR_073093.1 | 6 | 725 | ||||
| XM_011535894.1 | XP_011534196.1 | 5 | 1495 | |||
| XM_011535895.1 | XP_011534197.1 | 4 | 1279 | |||
| GenBank mRNA | AF125101.1 | - | 5 | 482 | ||
| AF151887.1 | - | 5 | 478 | |||
| AF275811.1 | - | 3 | 203 | |||
| AL556427.3 | - | 5 | 415 | |||
| AL561589.3 | - | 6 | 600 | |||
| BC000810.1 | - | 5 | 406 | 3452973 | ||
| BG828174.1 | - | 5 | 454 | |||

MPC1
Genomic Map