siRNA Details
| Gene Symbol: ZFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016107.3 | NP_057191.2 | 13 | 2305 | ||
| GenBank mRNA | AF100742.1 | - | 3 | 300 | ||
| AJ314790.1 | - | 13 | 2258 | |||
| BC051893.1 | - | 6 | 896 | |||
| BC062986.1 | - | 6 | 821 | |||
| BC137084.1 | - | 13 | 2269 | 9021081 | ||
| BC137085.1 | - | 13 | 2269 | 9021082 | ||

ZFR
Genomic Map