siRNA Details
| Gene Symbol: TELO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016111.3 | NP_057195.2 | 14 | 1973 | ||
| XM_011522773.2 | XP_011521075.1 | 14 | 1964 | |||
| XM_011522774.2 | XP_011521076.1 | 14 | 2162 | |||
| XM_011522775.2 | XP_011521077.1 | 14 | 1945 | |||
| XM_011522776.2 | XP_011521078.1 | 14 | 2032 | |||
| XM_011522777.2 | XP_011521079.1 | 14 | 1964 | |||
| XM_011522778.2 | XP_011521080.1 | 14 | 1964 | |||
| XM_017023914.1 | XP_016879403.1 | 14 | 1945 | |||
| GenBank mRNA | AB014583.3 | - | 14 | 1961 | ||
| AL080126.1 | - | 14 | 1904 | |||
| AL137394.1 | - | 14 | 1904 | |||
| BC017188.2 | - | 13 | 1729 | 3139743 | ||

TELO2
Genomic Map