siRNA Details
| Gene Symbol: NUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243351.1 | NP_001230280.1 | 3 | 294 | ||
| NM_016118.4 | NP_057202.3 | 3 | 294 | |||
| XM_005250011.2 | XP_005250068.1 | 3 | 359 | |||
| XM_017012304.1 | XP_016867793.1 | 3 | 356 | |||
| XM_017012305.1 | XP_016867794.1 | 3 | 356 | |||
| XM_017012306.1 | XP_016867795.1 | 3 | 375 | |||
| XM_017012307.1 | XP_016867796.1 | 3 | 356 | |||
| XM_017012308.1 | XP_016867797.1 | 3 | 356 | |||
| XM_017012309.1 | XP_016867798.1 | 3 | 359 | |||
| XM_017012310.1 | XP_016867799.1 | 3 | 355 | |||
| XM_017012311.1 | XP_016867800.1 | 3 | 259 | |||
| XM_017012312.1 | XP_016867801.1 | 3 | 231 | |||
| XM_017012313.1 | XP_016867802.1 | 3 | 355 | |||
| XM_017012314.1 | XP_016867803.1 | 3 | 231 | |||
| GenBank mRNA | AF155099.1 | - | 3 | 226 | ||
| AF300717.1 | - | 3 | 283 | |||
| AF459743.1 | - | 3 | 242 | |||
| AK293159.1 | - | 3 | 231 | |||
| AY129295.1 | - | 3 | 242 | |||
| BC046354.2 | - | 2 | 210 | 6070128 | ||
| DB048887.1 | - | 3 | 294 | |||

NUB1
Genomic Map