siRNA Details
| Gene Symbol: HSD17B12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016142.2 | NP_057226.1 | 4 | 604 | ||
| XM_011520156.1 | XP_011518458.1 | 4 | 598 | |||
| XM_017017881.1 | XP_016873370.1 | 4 | 284 | |||
| GenBank mRNA | AF078850.1 | - | 4 | 442 | ||
| AK027882.1 | - | 497 | ||||
| AK074952.1 | - | 4 | 456 | |||
| AK075011.1 | - | 4 | 489 | |||
| AK075216.1 | - | 4 | 465 | |||
| AK222881.1 | - | 4 | 426 | |||
| AK292625.1 | - | 4 | 489 | |||
| AK301663.1 | - | 5 | 395 | |||
| BC012043.1 | - | 4 | 427 | 3622879 | ||

HSD17B12
Genomic Map