siRNA Details
| Gene Symbol: WDR83OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016145.3 | NP_057229.1 | 4 | 591 | ||
| XM_017026864.1 | XP_016882353.1 | 5 | 1585 | |||
| GenBank mRNA | AF059620.1 | - | 4 | 359 | ||
| AF078861.1 | - | 4 | 576 | |||
| AF151898.1 | - | 4 | 575 | |||
| AK091030.1 | - | 4 | 2296 | |||
| BC001192.1 | - | 4 | 339 | |||
| BC042919.1 | - | 4 | 360 | 5932109 | ||
| BC061912.1 | - | 4 | 360 | 4444565 | ||

WDR83OS
Genomic Map