siRNA Details
| Gene Symbol: MMP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016155.5 | NP_057239.4 | 9 | 1321 | ||
| XM_011538355.2 | XP_011536657.1 | 9 | 1175 | |||
| XM_011538356.2 | XP_011536658.1 | 9 | 1171 | |||
| XM_011538357.2 | XP_011536659.1 | 9 | 1120 | |||
| GenBank mRNA | AB021225.2 | - | 9 | 1327 | ||
| BC040507.1 | - | 9 | 1313 | 5301089 | ||
| BC045610.1 | - | 9 | 1330 | 5273795 | ||
| X89576.1 | - | 8 | 1199 | |||

MMP17
Genomic Map