siRNA Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082.4 | NP_001073.3 | 6 | 656 | ||
| GenBank mRNA | AK296200.1 | - | 4 | 503 | ||
| AK300961.1 | - | 5 | 462 | |||
| BC067437.1 | - | 6 | 597 | 7002207 | ||
| BC067438.1 | - | 6 | 597 | |||
| BC067439.1 | - | 6 | 597 | 7002212 | ||
| BC067440.1 | - | 6 | 597 | 7002214 | ||
| D26480.1 | - | 6 | 601 | |||
| U02388.2 | - | 6 | 619 | |||

CYP4F2
Genomic Map