siRNA Details
| Gene Symbol: TELO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016111.3 | NP_057195.2 | 21 | 3225 | ||
| XM_011522773.2 | XP_011521075.1 | 22 | 3402 | |||
| XM_011522774.2 | XP_011521076.1 | 22 | 3600 | |||
| XM_011522775.2 | XP_011521077.1 | 22 | 3383 | |||
| XM_011522776.2 | XP_011521078.1 | 22 | 3470 | |||
| XM_017023914.1 | XP_016879403.1 | 21 | 3197 | |||
| GenBank mRNA | AB014583.3 | - | 21 | 3213 | ||
| AK001786.1 | - | 11 | 1661 | |||
| AL080126.1 | - | 21 | 3156 | |||
| AL137394.1 | - | 21 | 3156 | |||
| BC017188.2 | - | 20 | 2981 | 3139743 | ||
| BC028332.1 | - | 4 | 750 | |||

TELO2
Genomic Map