siRNA Details
| Gene Symbol: KCTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319294.1 | NP_001306223.1 | 18 | 2509 | ||
| NM_001319295.1 | NP_001306224.1 | 18 | 2581 | |||
| NM_016121.4 | NP_057205.2 | 18 | 2515 | |||
| XM_005273158.1 | XP_005273215.1 | 18 | 2781 | |||
| GenBank mRNA | AK296275.1 | - | 16 | 2048 | ||
| AK316272.1 | - | 14 | 1910 | |||
| AL833887.1 | - | 4 | 709 | |||
| AL834389.1 | - | 12 | 1739 | |||
| BC037896.1 | - | 2 | 1374 | |||
| BC094879.1 | - | 18 | 2458 | 3078971 | ||
| BC117188.1 | - | 18 | 2397 | 40125739 | ||
| BC126173.1 | - | 18 | 2397 | 8991889 | ||

KCTD3
Genomic Map