siRNA Details
| Gene Symbol: RNFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016125.3 | NP_057209.3 | 9 | 1427 | ||
| GenBank mRNA | AK098649.1 | - | 7 | 807 | ||
| BC006971.1 | - | 8 | 1304 | |||
| BC088365.1 | - | 9 | 1390 | 30390062 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNFT1| Gene Symbol: RNFT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016125.3 | NP_057209.3 | 9 | 1427 | ||
| GenBank mRNA | AK098649.1 | - | 7 | 807 | ||
| BC006971.1 | - | 8 | 1304 | |||
| BC088365.1 | - | 9 | 1390 | 30390062 | ||