siRNA Details
| Gene Symbol: SARAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB028926.1 | - | 6 | 1723 | ||
| AF078855.1 | - | 6 | 1691 | |||
| AF100748.1 | - | 6 | 1619 | |||
| AK025927.1 | - | 6 | 1773 | |||
| AK094386.1 | - | 7 | 1704 | |||
| AK130057.1 | - | 5 | 1471 | |||
| AY359104.1 | - | 6 | 1788 | |||
| BC015012.1 | - | 6 | 1687 | 3908758 | ||
| JQ348891.1 | - | 6 | 1732 | |||

SARAF
Genomic Map